Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MSAAMRERFD RFLHEKNCMT DLLAKLEAKT GVNRSFIALG VIGLVALYLV FGYGASLLCN LIGFGYPAYI SIKAIESPNK EDDTQWLTYW VVYGVFSIAE FFSDIFLSWF PFYYMLKCGF LLWCMAPSPS NGAELLYKRI IRPFFLKHES QMDSVVKDLK DKAKETADAI TKEAKKATVN LLGEEKKST
REEP5 (receptor expression-enhancing protein 5), also known as C5orf18, DP1, TB2 or D5S346, is a 189 amino acid multi-pass membrane protein. Thought to promote the functional cell surface expression of olfactory receptors, REEP5 belongs to the DP1 family and is encoded by a gene that maps to chromosome 5. With 181 million base pairs encoding around 1,000 genes, chromosome 5 is about 6% of human genomic DNA. Chromosome 5 is associated with Cockayne syndrome through the ERCC8 gene and familial adenomatous polyposis through the adenomatous polyposis coli (APC) tumor suppressor gene. Treacher Collins syndrome is also chromosome 5 associated and is caused by insertions or deletions within the TCOF1 gene. Deletion of the parm of chromosome 5 leads to cri du chat syndrome. Deletion of 5q or chromosome 5 altogether is common in therapy-related acute myelogenous leukemias and myelodysplastic syndrome.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: deleted in polyposis 1; polyposis coli region hypothetical protein DP1; Polyposis locus protein 1; Protein TB2; receptor expression enhancing protein 5; Receptor expression-enhancing protein 5
Gene Aliases: C5orf18; D5S346; DP1; REEP5; TB2; Yip2e; YOP1
UniProt ID: (Human) Q00765
Entrez Gene ID: (Human) 7905, (Pig) 100516959
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support