Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MRLSALLALA SKVTLPPHYR YGMSPPGSVA DKRKNPPWIR RRPVVVEPIS DEDWYLFCGD TVEILEGKDA GKQGKVVQVI RQRNWVVVGG LNTHYRYIGK TMDYRGTMIP SEAPLLHRQV KLVDPMDRKP TEIEWRFTEA GERVRVSTRS GRIIPKPEFP RADGIVPETW IDGPKDTSVE DALERTYVPC LKTLQEEVME AMGIKETRKY KKVYWY
Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 39S subunit protein which is more than twice the size of its E. coli counterpart. Sequence analysis identified two transcript variants that encode the same protein.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 39S ribosomal protein L24, mitochondrial; FLJ20917; L24mt; Large ribosomal subunit protein uL24m; MGC22737; MGC9831; MRPL24 protein
Gene Aliases: L24mt; MRP-L18; MRP-L24; MRPL24
UniProt ID: (Human) Q96A35
Entrez Gene ID: (Human) 79590
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support