Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Catalog Number | Type | Hinge Side | For Use With (Equipment) |
---|---|---|---|
6996TS also known as 6996 | Refrigerator and Freezer Door, Glass | Left | 11.5 cu. ft. (326L) Forma, Jewett, Puffer Hubbard, and Revco High Performance |
6995 | Refrigerator and Freezer Door, Solid | Left | 11.5 cu. ft. (326L) Forma, Jewett, Puffer Hubbard, and Revco High Performance |
6997 | Refrigerator and Freezer Door, Solid | Left | 23.3 cu. ft. (659L) Forma, Jewett, Puffer Hubbard, and Revco High Performance |
6998 | Refrigerator and Freezer Door, Glass | Left | 23.3 cu. ft. (659L) Forma, Jewett, Puffer Hubbard, and Revco High Performance |
6999TS also known as 6999 | Refrigerator and Freezer Door, Solid | Left | 29.2 cu. ft. (826L) Forma, Jewett, Puffer Hubbard, and Revco High Performance |
7001 | Refrigerator and Freezer Door, Solid | Left/Left | 45.8 cu. ft. (1297L) Forma, Jewett, Puffer Hubbard, and Revco High Performance |
7012TS also known as 7012 | Refrigerator and Freezer Door, Glass | Right | 23.3 cu. ft. (659L) Forma, Jewett, Puffer Hubbard, and Revco High Performance |
7014 | Refrigerator and Freezer Door, Glass | Left/Right | 45.8 cu. ft. (1297L) Forma, Jewett, Puffer Hubbard, and Revco High Performance |
7066 | Refrigerator and Freezer Door, Glass | Right | 11.5 cu. ft. (326L) Forma, Jewett, Puffer Hubbard, and Revco High Performance |
7067 | Refrigerator and Freezer Door, Glass | Left/Left | 45.8 cu. ft. (1297L) Forma, Jewett, Puffer Hubbard, and Revco High Performance |
7068 | Refrigerator and Freezer Door, Solid | Right/Right | 45.8 cu. ft. (1297L) Forma, Jewett, Puffer Hubbard, and Revco High Performance |
7069 | Refrigerator and Freezer Door, Glass | Right/Right | 45.8 cu. ft. (1297L) Forma, Jewett, Puffer Hubbard, and Revco High Performance |
7073 | Refrigerator and Freezer Door, Heated Glass | Left | 11.5 cu. ft. (326L) Forma, Jewett, Puffer Hubbard, and Revco High Performance |
7074 | Refrigerator and Freezer Door, Heated Glass | Right | 11.5 cu. ft. (326L) Forma, Jewett, Puffer Hubbard, and Revco High Performance |
7075TS also known as 7075 | Refrigerator and Freezer Door, Heated Glass | Left | 23.3 cu. ft. (659L) Forma, Jewett, Puffer Hubbard, and Revco High Performance |
7086 | Refrigerator and Freezer Door, Heated Glass | Right | 23.3 cu. ft. (659L) Forma, Jewett, Puffer Hubbard, and Revco High Performance |
7087 | Refrigerator and Freezer Door, Heated Glass | Left | 29.2 cu. ft. (826L) Forma, Jewett, Puffer Hubbard, and Revco High Performance |
7088 | Refrigerator and Freezer Door, Heated Glass | Right | 29.2 cu. ft. (826L) Forma, Jewett, Puffer Hubbard, and Revco High Performance |
7089 | Refrigerator and Freezer Door, Heated Glass | Left/Left | 45.8 cu. ft. (1297L) Forma, Jewett, Puffer Hubbard, and Revco High Performance |
7090 | Refrigerator and Freezer Door, Heated Glass | Right/Right | 45.8 cu. ft. (1297L) Jewett, Puffer Hubbard, and Revco High Performance |
7091 | Refrigerator and Freezer Door, Heated Glass | Left/Right | 45.8 cu. ft. (1297L) Forma, Jewett, Puffer Hubbard, and Revco High Performance |
7118 | Refrigerator and Freezer Door, Glass | Right | 4.7 cu. ft. (133L) Forma™, Jewett, Puffer Hubbard™ |
7119 | Refrigerator and Freezer Door, Solid | Left | 4.7 cu. ft. (133L) Forma™, Jewett, Puffer Hubbard™ |
7123 | Refrigerator and Freezer Door, Glass | Left | 4.7 cu. ft. (133L) Puffer Hubbard and Revco High Performance |
LE7013 | Refrigerator and Freezer Door, Glass | Right | 29.2 cu. ft. (826L) Forma, Jewett, Puffer Hubbard, and Revco High Performance |
For use with Thermo Scientific™ Forma™, Jewett™, Puffer Hubbard™ and Revco™ series. Solid doors are standard on most refrigerators unless otherwise specified. Optional dual-pane glass doors must be specified when ordering. Contact your Sales Representative for information about heated glass doors.
Ordering Information:
Door options must be specified at time of order