Thermo Scientific Chemicals

Indole-3-acetic acid, 98+%, Thermo Scientific Chemicals

Catalog number: A10556.36
500 g, Each
Thermo Scientific Chemicals

Indole-3-acetic acid, 98+%, Thermo Scientific Chemicals

Catalog number: A10556.36
500 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: A10556.36
also known as A10556-36
Price (EUR)
Quantity
-

Chemical Identifiers

CAS
87-51-4
IUPAC Name
2-(1H-indol-3-yl)acetic acid
Molecular Formula
C10H9NO2
InChI Key
SEOVTRFCIGRIMH-UHFFFAOYSA-N
SMILES
OC(=O)CC1=CNC2=CC=CC=C12
Identification (FTIR)
Conforms
Appearance (Color)
Pale cream to cream to pale brown or pale pink to gray
Form
Crystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)
≥97.5 to ≤102.5%
Assay (Silylated GC)
≥97.5%

Description

It is applied as plant growth hormone. is an inducer of plant cell elongation and division shown to cause uncontrolled growth.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is applied as plant growth hormone. is an inducer of plant cell elongation and division shown to cause uncontrolled growth.

Solubility
Soluble in ethanol (50 mg/ml), methanol, DMSO, and chloroform (sparingly). Insoluble in water.

Notes
Light sensitive. Store at -20°C. ncompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text

    Safety and Handling


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Skin Corrosion/Irritation
    Category 2
    Serious Eye Damage/Eye Irritation
    Category 2
    Specific target organ toxicity - (single exposure)
    Category 3
    Label Elements
    Signal Word

    Warning

    Hazard Statements

    H315 - Causes skin irritation

    H319 - Causes serious eye irritation

    H335 - May cause respiratory irritation

    Precautionary Statements

    P280 - Wear protective gloves/protective clothing/eye protection/face protection

    P302 + P352 - IF ON SKIN: Wash with plenty of soap and water

    P304 + P340 - IF INHALED: Remove person to fresh air and keep comfortable for breathing

    P305 + P351 + P338 - IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing

    P312 - Call a POISON CENTER or doctor if you feel unwell