Thermo Scientific Chemicals

2'-Deoxyadenosine monohydrate, 99%

Catalog number: A11166.14
25 g, Each
Thermo Scientific Chemicals

2'-Deoxyadenosine monohydrate, 99%

Catalog number: A11166.14
25 g, Each
Quantity
Catalog number: A11166.14
also known as A11166-14
Price (EUR)
Quantity
-

Chemical Identifiers

CAS
16373-93-6
IUPAC Name
(2R,3S,5R)-5-(6-amino-9H-purin-9-yl)-2-(hydroxymethyl)oxolan-3-ol
Molecular Formula
C10H13N5O3
InChI Key
OLXZPDWKRNYJJZ-YUZWJPFSNA-N
SMILES
NC1=C2N=CN([C@H]3C[C@H](O)[C@@H](CO)O3)C2=NC=N1
Water Content (Karl Fischer Titration)
5.0 to 8.0% (theory 6.69%)
Optical Rotation
-25.3? ± 1.5? (c = 0.5, Water)
Appearance (Color)
White
Assay (HPLC)
≥98.5%
Melting Point (clear melt)
188.0-194.0?C

Description

2?-Deoxyadenosineise is used by some cells as an energy source under energy stress conditions and to affect cAMP levels. 2?-dAdo is used in comparison studies of the functions of adenosine analogues on various biological processes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2′-Deoxyadenosineise is used by some cells as an energy source under energy stress conditions and to affect cAMP levels. 2′-dAdo is used in comparison studies of the functions of adenosine analogues on various biological processes.

Solubility
Soluble in water.

Notes
Keep container tightly closed. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text

    Safety and Handling


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Acute oral toxicity
    Category 4
    Label Elements
    Signal Word

    Warning

    Hazard Statements

    H302 - Harmful if swallowed

    Precautionary Statements

    P301 + P312 - IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell