Thermo Scientific Chemicals

Sorbic acid, 99%, Thermo Scientific Chemicals

Catalog number: A16196.36
500 g, Each
Thermo Scientific Chemicals

Sorbic acid, 99%, Thermo Scientific Chemicals

Catalog number: A16196.36
500 g, Each
Quantity
Catalog number: A16196.36
also known as A16196-36
Price (EUR)
Quantity
-

Chemical Identifiers

CAS
110-44-1
IUPAC Name
(2E,4E)-hexa-2,4-dienoic acid
Molecular Formula
C6H8O2
InChI Key
WSWCOQWTEOXDQX-MQQKCMAXSA-N
SMILES
C\C=C\C=C\C(O)=O
Appearance (Color)
White to pale cream
Form
Crystals or powder or crystalline powder or lumps
Assay (Silylated GC)
≥98.5%
Melting Point (clear melt)
131.0-137.0?C
Assay (Aqueous acid-base Titration)
≥98.5 to ≤101.5%

Description

Sorbic acid is used as a preservative and is found in food, cosmetics, and other household products. It acts as a stabilizers and antioxidants.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sorbic acid, is used as a food additive and has functional use such as antimicrobial preservatives, fungistatic agents. Sorbic acid is used as a preservative and is found in food, cosmetics, and other household products. It prevents the growth of mold, yeast, and fungi. It acts as a stabilizers and antioxidants.

Solubility
It is hardly soluble in water but soluble in glacial acetic acid, ethanol, propane diol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature 2 - 8°C. Keep away from Bases, Oxidizing agents, Reducing agentsBases, Oxidizing agents, Reducing agents. Photosensitive.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text

    Safety and Handling


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Skin Corrosion/Irritation
    Category 2
    Serious Eye Damage/Eye Irritation
    Category 2
    Specific target organ toxicity - (single exposure)
    Category 3
    Label Elements
    Signal Word

    Warning

    Hazard Statements

    H315 - Causes skin irritation

    H319 - Causes serious eye irritation

    H335 - May cause respiratory irritation

    Precautionary Statements

    P261 - Avoid breathing dust/fume/gas/mist/vapors/spray

    P280 - Wear protective gloves/protective clothing/eye protection/face protection

    P302 + P352 - IF ON SKIN: Wash with plenty of soap and water

    P305 + P351 + P338 - IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing