n-Propyl formate, 97%, Thermo Scientific Chemicals
n-Propyl formate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

n-Propyl formate, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
Catalog number A18090.AP
also known as A18090-AP
Price (EUR)/ Each
117,00
Online exclusive
130.00 
Save 13,00 (10%)
-
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (EUR)/ Each
117,00
Online exclusive
130.00 
Save 13,00 (10%)
Add to cart
n-Propyl formate, 97%, Thermo Scientific Chemicals
Catalog numberA18090.AP
Price (EUR)/ Each
117,00
Online exclusive
130.00 
Save 13,00 (10%)
-
Add to cart
Chemical Identifiers
CAS110-74-7
IUPAC Namepropyl formate
Molecular FormulaC4H8O2
InChI KeyKFNNIILCVOLYIR-UHFFFAOYSA-N
SMILESCCCOC=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥96.0%
Identification (FTIR)Conforms
Refractive Index1.3745-1.3795 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only