Search Thermo Fisher Scientific
图: 1 / 1
For reconstitution, we recommend adding 100 µL distilled water to a final antibody concentration of about 1 mg/mL. To use this carrier-free antibody for conjugation experiments, we strongly recommend performing another round of desalting. (Zeba Spin Desalting Columns, 7KMWCO, 0.5 mL, Product # 89882)
May be catalytically inactive.
仅用于科研。不用于诊断过程。未经明确授权不得转售。
蛋白别名: Deubiquitinating enzyme 40; Ubiquitin carboxyl-terminal hydrolase 40; ubiquitin specific protease 40; Ubiquitin thioesterase 40; ubiquitin thiolesterase 40; Ubiquitin-specific-processing protease 40
基因别名: USP40
UniProt ID: (Human) Q9NVE5
Entrez Gene ID: (Human) 55230