Thermo Scientific Chemicals

2,2-Bis[4-(4-aminophenoxy)phenyl]propane, 98%, Thermo Scientific Chemicals

Catalog number: H55057.14
25 g, Each
Thermo Scientific Chemicals

2,2-Bis[4-(4-aminophenoxy)phenyl]propane, 98%, Thermo Scientific Chemicals

Catalog number: H55057.14
25 g, Each
Quantity
Catalog number: H55057.14
also known as H55057-14
Price (EUR)
Quantity
-

Chemical Identifiers

CAS
13080-86-9
IUPAC Name
4-(4-{2-[4-(4-aminophenoxy)phenyl]propan-2-yl}phenoxy)aniline
Molecular Formula
C27H26N2O2
InChI Key
KMKWGXGSGPYISJ-UHFFFAOYSA-N
SMILES
CC(C)(C1=CC=C(OC2=CC=C(N)C=C2)C=C1)C1=CC=C(OC2=CC=C(N)C=C2)C=C1
Assay (HPLC)
>97.5%

Description

It is applied in pharmaceutical industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is applied in pharmaceutical industry.

Solubility
Very slightly soluble in water. Soluble in acetone, Dimethyl sulfoxide(DMSO).

Notes
Store at room temperature with proper ventilation. Keep away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text

    Safety and Handling


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Acute oral toxicity
    Category 4
    Skin Sensitization
    Category 1
    Chronic aquatic toxicity
    Category 4
    Label Elements
    Signal Word

    Warning

    Hazard Statements

    H302 - Harmful if swallowed

    H317 - May cause an allergic skin reaction

    H413 - May cause long lasting harmful effects to aquatic life

    Precautionary Statements

    P280 - Wear protective gloves/protective clothing

    P301 + P330 + P331 - IF SWALLOWED: rinse mouth. Do NOT induce vomiting

    P302 + P352 - IF ON SKIN: Wash with plenty of soap and water

    P312 - Call a POISON CENTER or doctor if you feel unwell

    P333 + P313 - If skin irritation or rash occurs: Get medical advice/attention