Phosphatase Inhibitor Cocktail I
Thermo Scientific Chemicals
Phosphatase Inhibitor Cocktail I
Have Questions?
Quantity:
5 mL
Catalog number J63907.AA
Price (EUR)/ Each
-
Quantity:
5 mL
Request bulk or custom format
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear
CommentComponents are dissolved in 18.2 megohm water, filtered through 0.22 micron filter
FormLiquid
Phosphatase Inhibitor Cocktail I is used to inhibitors with a broad specificity for serine, cysteine, and acid proteases, and aminopeptidases. It has been tested on a wide variety of cells and tissue extracts to isolate/maintain intact proteins without degradation.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Contents & Storage
-30°C to -10°C