Oxolinic acid, 98%, Thermo Scientific Chemicals
Oxolinic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Oxolinic acid, 98%, Thermo Scientific Chemicals

Targets gram-negative bacteria
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J66637.06
also known as J66637-06
Price (EUR)/ Each
84,02
Online exclusive
94.40 
Save 10,38 (11%)
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (EUR)/ Each
84,02
Online exclusive
94.40 
Save 10,38 (11%)
Add to cart
Oxolinic acid, 98%, Thermo Scientific Chemicals
Catalog numberJ66637.06
Price (EUR)/ Each
84,02
Online exclusive
94.40 
Save 10,38 (11%)
-
Add to cart
Specifications
Chemical Name or MaterialOxolinic acid
TypeOxolinic Acid
Physical FormPowder
CAS14698-29-4
Health Hazard 1H302
View more
Targets Gram-negative bacteria

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only