Thermo Scientific Chemicals

4-Methyl-1-pentyne, 97%, Thermo Scientific Chemicals

Catalog number: L05537.03
1 g, Each
Thermo Scientific Chemicals

4-Methyl-1-pentyne, 97%, Thermo Scientific Chemicals

Catalog number: L05537.03
1 g, Each
Quantity
Catalog number: L05537.03
also known as L05537-03
Price (EUR)
Quantity
-

Description

4-Methyl-1-pentyne, is used as a polymer matrix together with synthesized NH2-MIL 53 metal organic framework (MOF) as a filler were used to fabricate a mixed matrix membrane (MMM). It is also used in addition reactions which are typical in alkyne reactions such as halogenation, hydrogenation, hydrohalogenation, hydration, oxidative cleavage, nitrile formation and acidity of terminal alkynes. Polymerisation and substitution reactions are also useful in chemical synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methyl-1-pentyne, is used as a polymer matrix together with synthesized NH2-MIL 53 metal organic framework (MOF) as a filler were used to fabricate a mixed matrix membrane (MMM). It is also used in addition reactions which are typical in alkyne reactions such as halogenation, hydrogenation, hydrohalogenation, hydration, oxidative cleavage, nitrile formation and acidity of terminal alkynes. Polymerisation and substitution reactions are also useful in chemical synthesis.

Solubility
Insoluble in water. Soluble in common organic solvents of low polarity.

Notes
Stable under normal temperatures and pressures. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text

    Safety and Handling


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Flammable liquids
    Category 2
    Skin Corrosion/Irritation
    Category 2
    Serious Eye Damage/Eye Irritation
    Category 2
    Specific target organ toxicity - (single exposure)
    Category 3
    Label Elements
    Signal Word

    Danger

    Hazard Statements

    H315 - Causes skin irritation

    H319 - Causes serious eye irritation

    H335 - May cause respiratory irritation

    Physical Hazards

    H225 - Highly flammable liquid and vapor

    Precautionary Statements

    P261 - Avoid breathing dust/fume/gas/mist/vapors/spray

    P280 - Wear protective gloves/protective clothing/eye protection/face protection

    P302 + P352 - IF ON SKIN: Wash with plenty of soap and water

    P305 + P351 + P338 - IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing

    P210 - Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking

    P240 - Ground and bond container and receiving equipment