Thermo Scientific Chemicals

6-Heptenenitrile, 96%, Thermo Scientific Chemicals

Catalog number: L13137.06
5 g, Each
Thermo Scientific Chemicals

6-Heptenenitrile, 96%, Thermo Scientific Chemicals

Catalog number: L13137.06
5 g, Each
Quantity
Catalog number: L13137.06
also known as L13137-06
Price (EUR)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5048-25-9
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid or viscous liquid
Assay (GC)
≥96.0%
Refractive Index
1.4305-1.4355 @ 20?C

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text

    Safety and Handling


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Acute oral toxicity
    Category 4
    Acute dermal toxicity
    Category 4
    Acute Inhalation Toxicity - Vapors
    Category 4
    Label Elements
    Signal Word

    Warning

    Hazard Statements

    H302 + H312 + H332 - Harmful if swallowed, in contact with skin or if inhaled

    Precautionary Statements

    P280 - Wear protective gloves/protective clothing

    P301 + P330 + P331 - IF SWALLOWED: rinse mouth. Do NOT induce vomiting

    P302 + P352 - IF ON SKIN: Wash with plenty of soap and water

    P304 + P340 - IF INHALED: Remove person to fresh air and keep comfortable for breathing

    P312 - Call a POISON CENTER or doctor if you feel unwell