Thermo Scientific Chemicals

1-Ethylpiperazine, 98%, Thermo Scientific Chemicals

Catalog number: L15539.22
100 g, Each
Thermo Scientific Chemicals

1-Ethylpiperazine, 98%, Thermo Scientific Chemicals

Catalog number: L15539.22
100 g, Each
Quantity
Catalog number: L15539.22
also known as L15539-22
Price (EUR)
Quantity
-

Chemical Identifiers

CAS
5308-25-8
IUPAC Name
1-ethylpiperazine
Molecular Formula
C6H14N2
InChI Key
WGCYRFWNGRMRJA-UHFFFAOYSA-N
SMILES
CCN1CCNCC1
Identification (FTIR)
Conforms
Assay (GC)
≥97.5%
Refractive Index
1.4670-1.4710 @ 20?C
Appearance (Color)
Clear colorless to yellow
Form
Liquid

Description

1-Ethylpiperazine is used in the synthesis of 2-(2-methoxy-5-((4-methylpiperazin-1-yl)sulfonyl)phenyl)-1H-benzo[d]imidazole hydrochloride and 2-(5-((4-ethylpiperazin-1-yl)sulfonyl)-2-methoxyphenyl)-1H-benzo[d]imidazole hydrochloride. It is also employed as an intermediate in veterinary medicine, especially in the production of ciproflmoxacin ethyl. Further, it serves as a precursor to prepare dyes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Ethylpiperazine is used in the synthesis of 2-(2-methoxy-5-((4-methylpiperazin-1-yl)sulfonyl)phenyl)-1H-benzo[d]imidazole hydrochloride and 2-(5-((4-ethylpiperazin-1-yl)sulfonyl)-2-methoxyphenyl)-1H-benzo[d]imidazole hydrochloride. It is also employed as an intermediate in veterinary medicine, especially in the production of ciproflmoxacin ethyl. Further, it serves as a precursor to prepare dyes.

Solubility
Miscible with water.

Notes
Air sensitive. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text

    Safety and Handling


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Flammable liquids
    Category 3
    Acute oral toxicity
    Category 4
    Acute Inhalation Toxicity - Dusts and Mists
    Category 4
    Skin Corrosion/Irritation
    Category 1
    Serious Eye Damage/Eye Irritation
    Category 1
    Label Elements
    Signal Word

    Danger

    Hazard Statements

    H302 + H332 - Harmful if swallowed or if inhaled

    H314 - Causes severe skin burns and eye damage

    Physical Hazards

    H226 - Flammable liquid and vapor

    Precautionary Statements

    P301 + P312 - IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell

    P304 + P340 - IF INHALED: Remove person to fresh air and keep comfortable for breathing

    P210 - Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking

    P280 - Wear eye protection/ face protection

    P301 + P330 + P331 - IF SWALLOWED: rinse mouth. Do NOT induce vomiting

    P305 + P351 + P338 - IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing

    P310 - Immediately call a POISON CENTER or doctor/physician