[年中大促] 买1瓶巴西血清任选9瓶培养基,多套价优,最高立减4000元!

立即参与

Hamburger Menu Button
Thermo Fisher Scientific Logo
登录
没有账号?建立账号 注册
  • 产品
    • 抗体
    • 引物合成
    • GeneArt Gene Synthesis Instant Designer
    • TaqMan Assay and Array Search
    • PCR 设备和耗材
    • 色谱柱和纯化柱
    • 实验室离心机
    • 分子生物学试剂和试剂盒
    • 实验室塑料耗材及用品
    • 查看所有产品分类
  • 应用和技术
    • 色谱
    • 细胞转染和培养
    • 实时荧光定量PCR
    • 质谱
    • 实验室塑料耗材及用品
    • 电子显微镜
    • 蛋白质生物学
    • PCR
    • RNAi
    • 环境
    • 查看所有应用和技术
  • 服务
    • 定制服务
    • 企业级实验室信息化服务
    • 企业服务
    • 金融&租赁服务
    • 仪器服务
    • OEM和商业供应
    • 培训服务
    • 360° CDMO 和 CRO 服务
    • CDMO 服务
    • 临床试验 CRO 服务
    • 查看所有帮助和支持
  • 帮助和支持
    • 学习中心
    • 联系我们
    • 更改国家或地区
    • 查看所有帮助和支持
  • 热门产品
    • TaqMan Real-Time PCR Assays
      TaqMan 实时荧光定量 PCR 试剂盒
    • Antibodies
      抗体
    • Oligos, Primers & Probes
      寡核苷酸、引物、探针
    • GeneArt Gene Synthesis
      GeneArt 基因合成
    • Cell Culture Plastics
      细胞培养耗材
  • 联系我们
  • 货号直购
  • 订单查询
  • 查找文件&证书
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

一抗
  • 搜索全部或下列指定分类
  • Popular Product Areas
  • TaqMan试剂盒
  • 基因表达
  • microbe 和 ABR 检测
  • 一抗
  • 二抗
  • 同型对照
  • 蛋白质和多肽
  • ELISA 试剂盒
  • SNP基因分型
  • 所有文档和支持
  • 证书(COA)
  • 安全数据表 (SDS)
  • 产品常见问题
  • 说明书
    Search button Close
            • 联系我们
            • 货号直购
            • 登录
              登录
              没有账号?建立账号 注册
              • 我的账号
              • 订单
              • 账单及发票查询
              • 定制产品及项目

            Disclaimer

            Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

            Loading ...
            • Primary Antibodies ›
            • APBA1 Antibodies

            Invitrogen

            APBA1 Polyclonal Antibody

            查看其他6个APBA1抗体
            说明书
            实验方案
            问题与答案
            技术支持
            说明书
            实验方案
            问题与答案
            技术支持

            Cite APBA1 Polyclonal Antibody

            • 抗体检测数据 (1)
            Application
            APBA1 Antibody in Western Blot (WB)
            Group 53 Created with Sketch.

            图: 1 / 1

            APBA1 Antibody (PA5-18702) in WB

            Western blot analysis of APBA1 using APBA1 Polyclonal Antibody (Product # PA5-18702) (0.1 µg/mL) in staining of Human Brain (35 µg protein in RIPA buffer). Primary incubation was 1 hour. Detected by chemiluminescence.
            查看产品

            APBA1 Antibody in Western Blot (WB)

            产品信息

            PA5-18702

            应用
            建议稀释比
            已发表文章

            免疫印迹 (WB)

            0.1-0.3 µg/mL
            -
            产品规格

            种属反应

            Human

            宿主/亚型

            Goat / IgG

            分类

            Polyclonal

            类型

            Antibody

            抗原

            Synthetic peptide sequence (EASHPSQDGKRQYK) corresponding to the internal amino acids of APBA1 (aa 573-587).
            查看免疫原

            偶联物

            Unconjugated

            形式

            Liquid

            浓度

            0.5 mg/mL

            纯化类型

            Ammonium sulfate precipitation

            保存液

            TBS, pH 7.3, with 0.5% BSA

            内含物

            0.02% sodium azide

            保存条件

            -20°C, Avoid Freeze/Thaw Cycles

            运输条件

            Ambient (domestic); Wet ice (international)

            RRID

            AB_10987127

            产品详细信息

            This antibody is predicted to react with mouse and rat based on sequence homology.

            This antibody is tested in Peptide ELISA: antibody detection limit dilution 16,000.

            靶标信息

            The munc-18 interacting protein (Mint) protein family is a group of evolutionarily conserved adaptor proteins that function in membrane transport and organization. In mammals, there exist three Mint isoforms, Mint1, 2, and 3. Although there is little amino acid sequence conservation in the amino-terminal half, the carboxy-terminal half of these proteins is highly conserved. Within this conserved portion there exists a phosphotyrosine-binding (PTB) and a PSD-95/DLG-A/ZO-1 (PDZ) domain, which function as protein interaction modules. Mint1 and 2 appear to be expressed exclusively in the brain and are found to bind to Munc18, an essential component of the synaptic vesicle fusion machinery. Mint3 is ubiquitously expressed in all tissues and is expressed at the lowest levels in the brain and testis. Studies show that mint3 does not interact with munc-18. Mint3 has been found to interact with the Alzheimer's Disease-related amyloid precursor protein (APP) and does so through its PTB and PDZ domains. It has been suggested that mint3 links APP to other transport machinery components, thereby regulating it transport, endocytosis, and metabolism. Abnormal APP metabolism has been shown to be the cause of an early-onset type of Alzheimer's disease.

            仅用于科研。不用于诊断过程。未经明确授权不得转售。

            篇参考文献 (0)

            您是否在文献中引用过该产品?请点击下方按钮邮件告知我们。
            引用该产品

            生物信息学

            蛋白别名: Adapter protein X11alpha; adaptor protein X11alpha; amyloid beta (A4) precursor protein-binding, family A, member 1 (X11); Amyloid-beta A4 precursor protein-binding family A member 1; Mint-1; Neuron-specific X11 protein; Neuronal Munc18-1-interacting protein 1; phosphotyrosine-binding/-interacting domain (PTB)-bearing protein

            查看更多 收起

            基因别名: APBA1; D9S411E; LIN10; MINT1; X11; X11A; X11ALPHA

            查看更多 收起

            UniProt ID: (Human) Q02410

            查看更多 收起

            Entrez Gene ID: (Human) 320

            查看更多 收起

            功能

            beta-amyloid binding protein binding phosphatidylinositol-4,5-bisphosphate binding PDZ domain binding protein complex binding membrane trafficking regulatory protein

            参与通路

            in utero embryonic development protein complex assembly intracellular protein transport cell adhesion chemical synaptic transmission neurotransmitter secretion nervous system development locomotory behavior axo-dendritic transport regulation of gene expression glutamate secretion gamma-aminobutyric acid secretion multicellular organism growth
            It has to be done as per old AB suggested Products section.
            guarantee_icon

            性能保障

            只要您发现Invitrogen抗体在实验中的表现与网站或说明书的所述内容不符,您即可享受免费退换货服务。*

            了解更多
            help_icon

            随时为您服务

            获取常见问题的专家解答,或者直接联系我们的技术专家,咨询应用、设备、常规产品使用问题。

            联系技术支持
            订购 Plus Icon Minus Icon
            • 订单状态查询
            • 订单支持
            • 货号直购
            • 现货供应中心
            • 电子采购
            支持 Plus Icon Minus Icon
            • 帮助&支持
            • 联系我们 - 400 820 8982
            • 技术支持中心
            • 查找文件&证书
            • 报告网站问题
            资源 Plus Icon Minus Icon
            • 学习中心
            • 促销
            • 活动&研讨会
            • 社交媒体
            关于赛默飞 Plus Icon Minus Icon
            • 关于我们
            • 招聘
            • 投资者关系
            • 新闻
            • 社会责任
            • 商标
            品牌 Plus Icon Minus Icon
            • Thermo Scientific
            • Applied Biosystems
            • Invitrogen
            • Gibco
            • Ion Torrent
            • Patheon
            • PPD
            • 服务条款
            • 隐私政策
            • 定价和运输保险政策
            • Cookie 喜好设置

              Cookie设置

              我们及我们的关联方等通过运用Cookie等技术来为您提供您感兴趣的网站定制内容、识别访问者、保障安全登录和收集数据。点击“全部接受”以接受所有 cookie 并直接跳转至网站,或点击“管理设置”以查看 cookie 类型的详细说明,并选择是否在网站上接受某些 cookie。
            © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
            China flag icon
            China
            本网站销售的所有产品均不得用于人类或动物之临床诊断或治疗,仅可用于工业或者科研等非医疗目的。
               电子营业执照 | 经营证照公示 | ICP备案号:沪ICP备11003924号 | 沪公网安备 31011502006935号

            Your items have has been added!


            Host server : magellan-search-green-56756dfb8b-mfpfg:80/100.66.79.199:80.
            git-commit: de6905f249d25e22729e613aef254bb544bd8b7c
            git-url: https://github.com/thermofisher/magellan-search
            git-branch: release/2.31.1-2025.06.25-1.0