XLD Agar (EP, USP, JP, BP)
XLD Agar (EP, USP, JP, BP)
Thermo Scientific™

XLD Agar (EP, USP, JP, BP)

Isolate and identify Salmonella and Shigella spp. with this ready-to-use medium.

Have Questions?
Catalog number PO1132A
Price (EUR)
-
Request A Quote
Ask our AI about this Product

Isolate and identify Salmonella and Shigella spp. with ready-to-use Thermo Scientific™ Oxoid™ XLD Agar (EP, USP, JP, BP). The medium relies on xylose fermentation, lysine decarboxylation and production of hydrogen sulphide for the primary differentiation of shigellae and salmonellae from non-pathogenic bacteria. The careful addition of Sodium desoxycholate inhibits growth of coliforms without decreasing the ability to support shigellae and salmonellae. The medium conforms to the formulations detailed in international pharmacopoeia2,3,4,5.

Salmonella and Shigella are major causes of bacterial enteric illness with transmission occuring via the fecal-oral route, person-to-person contact, or by the ingestion of contaminated food or water.

Use XLD Agar (EP, USP, JP, BP) for the screening of samples containing mixed flora and suspected of harbouring enteric pathogens, e.g. food products.

  • Easy-to-read: easily differentiate Salmonella (red colonies with black centres) from Shigella (red colonies) and other coliforms (yellow, opaque colonies).
  • Superior sensitivity and selectivity: exceeds that of the traditional plating media e.g., Eosin Methylene Blue and Bismuth Sulphite Agars, which tend to suppress the growth of shigellae.
  • Tested in accordance with ISOL 1133:20141
  • Conforms to formulations detailed in International Pharmacopoeia2,3,4,5.
  • Ready-to-use convenience of prepared media.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

1. ISO 11133:2014 Microbiology of food, animal feed and water - Preparation, production, storage and performance testing of culture media.

2.British Pharmacopoeia 2005

3.European Pharmacopoeia 6th Edition 2008

4.The Japanese Pharmacopoeia JP 15 2006

5.The United States Pharmacopoeia USP 32 2009

Specifications
ColorRed, clear
DescriptionXLD Agar (EP, USP, JP, BP)
Fill Weight19.5g ± 1.0g
Format90mm Plate
Packaging TypeFlow Wrapped - Single Layer
Quantity10/Pk.
Shelf Life9 Weeks
Manufacturing OriginUnited Kingdom
Product TypeAgar
Unit SizeEach
Contents & Storage
2°C to 10°C
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Share catalog number, name or link

1x1 image pixel for data collection