Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Catalog Number | Quantity | Diameter (Metric) | Surface Acid Loading | Concentration |
---|---|---|---|---|
W090CB also known as MCG-W090CB-EACH | 100 mL | 0.9 μm | High Acid | 4% |
73003305100250 | 15 mL | 2 μm | High Acid | 10% |
73003305100350 | 100 mL | 2 μm | High Acid | 10% |
73003305100450 | 1000 mL | 2 μm | High Acid | 10% |
73003420100250 also known as MCG-73003420100250-EACH | 15 mL | 3 μm | High Acid | 10% |
73003420100350 | 100 mL | 3 μm | High Acid | 10% |
83000520100290 also known as MCG-83000520100290-EACH | 15 mL | 0.2 μm | High Acid | 10% |
83000520100390 | 100 mL | 0.2 μm | High Acid | 10% |
83000520100490 | 1000 mL | 0.2 μm | High Acid | 10% |
83000550100290 | 15 mL | 0.2 μm | Medium Acid | 10% |
83000550100390 | 100 mL | 0.2 μm | Medium Acid | 10% |
83000550100490 | 1000 mL | 0.2 μm | Medium Acid | 10% |
83000720100290 also known as MCG-83000720100290-EACH | 15 mL | 0.3 μm | High Acid | 10% |
83000720100390 | 100 mL | 0.3 μm | High Acid | 10% |
83000750100290 | 15 mL | 0.3 μm | Medium Acid | 10% |
83000750100390 | 100 mL | 0.3 μm | Medium Acid | 10% |
83000920100290 | 15 mL | 0.4 μm | High Acid | 10% |
83000920100390 | 100 mL | 0.4 μm | High Acid | 10% |
83000970100290 | 15 mL | 0.4 μm | Low Acid | 10% |
83000970100390 | 100 mL | 0.4 μm | Low Acid | 10% |
83000970100490 | 1000 mL | 0.4 μm | Low Acid | 10% |
93000570100290 also known as MCG-93000570100290-EACH | 15 mL | 0.2 μm | Low Acid | 10% |
93000570100390 | 100 mL | 0.2 μm | Low Acid | 10% |
93001891100290 | 15 mL | 0.85 μm | Low Acid | 10% |
93001891100390 | 100 mL | 0.85 μm | Low Acid | 10% |
W004CA | 15 mL | 0.4 μm | Low Acid | 4% |
W004CB | 100 mL | 0.4 μm | Low Acid | 4% |
W050CA | 15 mL | 0.5 μm | Medium Acid | 4% |
W050CB | 100 mL | 0.5 μm | Medium Acid | 4% |
W080CA | 15 mL | 0.85 μm | Medium Acid | 4% |
W080CB | 100 mL | 0.85 μm | Medium Acid | 4% |
W090CA also known as MCG-W090CA-EACH | 15 mL | 0.9 μm | High Acid | 4% |
W400CA | 15 mL | 4.0 μm | High Acid | 4% |
W400CB | 100 mL | 4.0 μm | High Acid | 4% |
W500CA | 15 mL | 5.0 μm | High Acid | 4% |
W500CB | 100 mL | 5.0 μm | High Acid | 4% |
Parking Area Properties
Recommended for:
Note:
When using Opti-Link particles, ask about our Microparticle Reagent Optimization manual, which outlines our reagent development strategy. This manual includes validated binding, coupling and particle reagent optimization protocols.