Thermo Scientific Chemicals

Barium chloride dihydrate, Puratronic™, 99.997% (metals basis), Thermo Scientific Chemicals

Catalog number: 010646.22
100 g, Each
Thermo Scientific Chemicals

Barium chloride dihydrate, Puratronic™, 99.997% (metals basis), Thermo Scientific Chemicals

Catalog number: 010646.22
100 g, Each
Quantity
Catalog number: 010646.22
also known as 010646-22
Price (EUR)
Quantity
-

Chemical Identifiers

CAS
10326-27-9
IUPAC Name
barium(2+) dihydrate dichloride
Molecular Formula
BaCl2H4O2
InChI Key
PWHCIQQGOQTFAE-UHFFFAOYSA-L
SMILES
O.O.[Cl-].[Cl-].[Ba++]
Total Metal Impurities
0.001% max.

Description

It is mainly used in the purification of brine solution in caustic chlorine plants, as a reagent in the test for sulfate ion in chemical lab, and as a cleansing agent in the production of other barium chemicals and lubricating oil additives. It is used in the production of molecular sieves, pigments, and paper coatings, and serves as a precursor to fabricate the doped barium zirconate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text

    Safety and Handling


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Acute oral toxicity
    Category 3
    Acute Inhalation Toxicity - Dusts and Mists
    Category 4
    Serious Eye Damage/Eye Irritation
    Category 2
    Label Elements
    Signal Word

    Danger

    Hazard Statements

    H301 - Toxic if swallowed

    H319 - Causes serious eye irritation

    H332 - Harmful if inhaled

    Precautionary Statements

    P280 - Wear eye protection/ face protection

    P301 + P310 - IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician

    P304 + P340 - IF INHALED: Remove person to fresh air and keep comfortable for breathing

    P337 + P313 - If eye irritation persists: Get medical advice/attention