Gold wire, 1.0mm (0.04in) dia, Premion™, 99.9985% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Gold wire, 1.0mm (0.04in) dia, Premion™, 99.9985% (metals basis), Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 cm
2 cm
50 cm
Catalog number 010965.BS
also known as 010965-BS
Price (EUR)/ Each
376,20
Online exclusive
418.00 
Save 41,80 (10%)
-
Add to cart
Quantity:
10 cm
Request bulk or custom format
Price (EUR)/ Each
376,20
Online exclusive
418.00 
Save 41,80 (10%)
Add to cart
Gold wire, 1.0mm (0.04in) dia, Premion™, 99.9985% (metals basis), Thermo Scientific Chemicals
Catalog number010965.BS
Price (EUR)/ Each
376,20
Online exclusive
418.00 
Save 41,80 (10%)
-
Add to cart
Chemical Identifiers
CAS7440-57-5
IUPAC Namegold
Molecular FormulaAu
InChI KeyPCHJSUWPFVWCPO-UHFFFAOYSA-N
SMILES[Au]
View more
SpecificationsSpecification SheetSpecification Sheet
FormWire, 1.0mm (0.04in) diameter
Assay from Supplier's CofA99.9985% (metals basis)
Appearance (Color)Golden
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only