Thermo Scientific Chemicals

Platinum(IV) oxide monohydrate, Adams Catalyst, Pt 75% min, Thermo Scientific Chemicals

Catalog number: 012278.03
1 g, Each
Thermo Scientific Chemicals

Platinum(IV) oxide monohydrate, Adams Catalyst, Pt 75% min, Thermo Scientific Chemicals

Catalog number: 012278.03
1 g, Each
Quantity
Catalog number: 012278.03
also known as 012278-03
Price (EUR)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
12137-21-2
IUPAC Name
dioxoplatinum hydrate
Molecular Formula
H2O3Pt
InChI Key
SPGAMGILENUIOF-UHFFFAOYSA-N
SMILES
O.O=[Pt]=O
Surface Area
85 m²/g min
Assay (unspecified)
Pt 75% min

Description

Hydrogenation catalyst. Suitable for the reduction of double and triple bonds, aromatics, carbonyls, nitriles, and nitro groupsPlatinum(IV) oxide monohydrate is used as a catalyst in hydrogenation, hydrogenolysis, dehydrogenation, and oxidation reactions in organic synthesis. It also finds application in ceramic industry, inorganic chemistry and in chemical synthesis. It is utilized for the selective reduction of alkenes without reducing nitro group.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Hydrogenation catalyst
  • Suitable for the reduction of double and triple bonds, aromatics, carbonyls, nitriles, and nitro groups
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text

    Safety and Handling


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Oxidizing solids
    Category 1
    Label Elements
    Signal Word

    Danger

    Hazard Statements

    H271 - May cause fire or explosion; strong oxidizer

    Precautionary Statements

    P210 - Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking

    P221 - Take any precaution to avoid mixing with combustibles

    P280 - Wear protective gloves/protective clothing/eye protection/face protection

    P371 + P380 + P375 - In case of major fire and large quantities: Evacuate area. Fight fire remotely due to the risk of explosion