Cesium perchlorate, Reagent Grade, Thermo Scientific Chemicals
Cesium perchlorate, Reagent Grade, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cesium perchlorate, Reagent Grade, Thermo Scientific Chemicals

Have Questions?
Quantity:
25 g
Catalog number 012308.14
also known as 012308-14
Price (EUR)/ Each
85,77
Online exclusive
95.30 
Save 9,53 (10%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (EUR)/ Each
85,77
Online exclusive
95.30 
Save 9,53 (10%)
Add to cart
Cesium perchlorate, Reagent Grade, Thermo Scientific Chemicals
Catalog number012308.14
Price (EUR)/ Each
85,77
Online exclusive
95.30 
Save 9,53 (10%)
-
Add to cart
Chemical Identifiers
CAS13454-84-7
IUPAC Namecaesium(1+) perchlorate
Molecular FormulaClCsO4
InChI KeyWKDKOOITVYKILI-UHFFFAOYSA-M
SMILES[Cs+].[O-][Cl](=O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Impurity content0.1% max
Cesium perchlorate is used for long-duration optical transmission of information with infofuses. It is a modifier of polymer electrolyte/luminescent conjugated polymer mixed ionic-electronic conductors. It has been used as a reactant for investigating metal salt effects on morphology of silica nanomaterials prepared by polymerization on hydrogels.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only