Thermo Scientific Chemicals

Barium zirconium oxide, 99% (metals basis), Thermo Scientific Chemicals

Catalog number: 014028.A1
1 kg, Each
Thermo Scientific Chemicals

Barium zirconium oxide, 99% (metals basis), Thermo Scientific Chemicals

Catalog number: 014028.A1
1 kg, Each
Quantity

Chemical Identifiers

CAS
12009-21-1
IUPAC Name
zirconium(4+) barium(2+) trioxidandiide
Molecular Formula
BaO3Zr
InChI Key
QLFKDQYBEKOEOT-UHFFFAOYSA-N
SMILES
[O--].[O--].[O--].[Zr+4].[Ba++]
Form
Powder
Assay (special)
Fe₂O₃: Report
Assay (special)
Na₂O: Report
Assay (special)
SiO₂: Report
Assay (special)
SrO: Report

Description

Barium zirconium oxide is involved in the preparation of its nano powder, which finds application in the gas sensing performance of thick films especially with ammonia gas. Copper(II) oxide doping with yttrium-doped barium zirconate is used as an electrolyte in solid oxide fuel cell.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text

    Safety and Handling


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Acute oral toxicity
    Category 4
    Acute Inhalation Toxicity - Dusts and Mists
    Category 4
    Label Elements
    Signal Word

    Warning

    Hazard Statements

    H302 + H332 - Harmful if swallowed or if inhaled

    Precautionary Statements

    P261 - Avoid breathing dust/fume/gas/mist/vapors/spray

    P264 - Wash face, hands and any exposed skin thoroughly after handling

    P301 + P330 + P331 - IF SWALLOWED: rinse mouth. Do NOT induce vomiting

    P304 + P340 - IF INHALED: Remove person to fresh air and keep comfortable for breathing

    P312 - Call a POISON CENTER or doctor if you feel unwell