Thermo Scientific Chemicals

Aluminum titanium oxide, 99.5% (metals basis), Thermo Scientific Chemicals

Catalog number: 014484.22
100 g, Each
Thermo Scientific Chemicals

Aluminum titanium oxide, 99.5% (metals basis), Thermo Scientific Chemicals

Catalog number: 014484.22
100 g, Each
Quantity
Catalog number: 014484.22
also known as 014484-22
Price (EUR)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
12004-39-6
IUPAC Name
dititanium(4+) dialuminium(3+) heptaoxidandiide
Molecular Formula
Al2O7Ti2
InChI Key
COTUMVFAJGQVPQ-UHFFFAOYSA-N
SMILES
[O--].[O--].[O--].[O--].[O--].[O--].[O--].[Al+3].[Al+3].[Ti+4].[Ti+4]
Total Metal Impurities
0.5% max.

Description

Aluminum titanium oxide is used in refractory industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text