Gold(I) iodide, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Gold(I) iodide, 99%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 016617.03
also known as 016617-03
Price (EUR)/ Each
176,40
Online exclusive
196.00 
Save 19,60 (10%)
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (EUR)/ Each
176,40
Online exclusive
196.00 
Save 19,60 (10%)
Add to cart
Gold(I) iodide, 99%, Thermo Scientific Chemicals
Catalog number016617.03
Price (EUR)/ Each
176,40
Online exclusive
196.00 
Save 19,60 (10%)
-
Add to cart
Chemical Identifiers
CAS10294-31-2
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)98.6% min
Gold(I) iodide is employed in the synthesis of chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only