Gallium Tin alloy, 99.99% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Gallium Tin alloy, 99.99% (metals basis), Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number 018161.09
also known as 018161-09
Price (EUR)/ Each
99,90
Online exclusive
111.00 
Save 11,10 (10%)
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (EUR)/ Each
99,90
Online exclusive
111.00 
Save 11,10 (10%)
Add to cart
Gallium Tin alloy, 99.99% (metals basis), Thermo Scientific Chemicals
Catalog number018161.09
Price (EUR)/ Each
99,90
Online exclusive
111.00 
Save 11,10 (10%)
-
Add to cart
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Silver
FormLow melting solid
Assay from Supplier's CofA≥99.99% (metals basis)
Assay from Supplier's CofASn:Ga; 8.0:92.0 wt%
Gallium Tin alloy is used to cool computer chips in place of water.  Used in medical thermometers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only