Multi Ion anion IC standard solution, Specpure™ (Waters) in H{2}O, Thermo Scientific Chemicals
Multi Ion anion IC standard solution, Specpure™ (Waters) in H{2}O, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Multi Ion anion IC standard solution, Specpure™ (Waters) in H{2}O, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
100 mL
Catalog number 035564.AE
also known as 035564-AE
Price (EUR)/ Each
228,60
Online exclusive
254.00 
Save 25,40 (10%)
-
Add to cart
Quantity:
100 mL
Request bulk or custom format
Price (EUR)/ Each
228,60
Online exclusive
254.00 
Save 25,40 (10%)
Add to cart
Multi Ion anion IC standard solution, Specpure™ (Waters) in H{2}O, Thermo Scientific Chemicals
Catalog number035564.AE
Price (EUR)/ Each
228,60
Online exclusive
254.00 
Save 25,40 (10%)
-
Add to cart
Specifications
Chemical Name or MaterialMulti Ion anion IC standard solution
Name NoteSpecpure™ (Waters) in H2O
Quantity100 mL
Recommended StorageAmbient temperatures
EINECS Number231-791-2
View more
Multi Ion anion IC standard solution is a high purity starting material, which is a certified primary reference material for Ion Chromatography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only