Aluminum Magnesium gauze, alloy 5056, 30 mesh woven from 0.23mm (0.009in) dia wire
Aluminum Magnesium gauze, alloy 5056, 30 mesh woven from 0.23mm (0.009in) dia wire
Aluminum Magnesium gauze, alloy 5056, 30 mesh woven from 0.23mm (0.009in) dia wire
Aluminum Magnesium gauze, alloy 5056, 30 mesh woven from 0.23mm (0.009in) dia wire
Thermo Scientific Chemicals

Aluminum Magnesium gauze, alloy 5056, 30 mesh woven from 0.23mm (0.009in) dia wire

Have Questions?
Change viewbuttonViewtableView
Quantity:
150 x 150 mm
75 x 75 mm
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 040906.GT
also known as 040906-GT
Price (EUR)
-
Quantity:
150 x 150 mm
Request bulk or custom format
Specifications
Avg. Mol. Wt. or Mol. Wt. RangeAl:Mg; 95:5 wt%
Chemical Name or MaterialAluminum Magnesium gauze, Alloy 5056
MDL NumberMFCD00214039
Mesh Size30 Mesh
Name NoteWoven from 0.23mm (0.009 in.) dia. wire
View more
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
Y25K050Certificate of AnalysisDec 11, 2024040906.GT, 040906.FL
W10K004Certificate of AnalysisSep 25, 2024040906.GT, 040906.FL
Q10E017Certificate of AnalysisDec 07, 2021040906.GT, 040906.FL
S10F008Certificate of AnalysisNov 23, 2021040906.GT, 040906.FL
L19K21Certificate of AnalysisNov 23, 2021040906.GT, 040906.FL
5 results displayed, search above for a specific certificate

Safety Data Sheets

Safety and Handling

EU Specific Hazard Statements

EUH210 - Safety data sheet available on request

Share catalog number, name or link

1x1 image pixel for data collection