Glassy carbon plate, 3mm (0.1 in.) thick, type 2, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Glassy carbon plate, 3mm (0.1 in.) thick, type 2, Thermo Scientific Chemicals
Used in casting, smelting of metal, hard alloys. Glassy carbon is widely used as an electrode material in electrochemistry, for high temperature crucibles and as a part of some prosthetic devices.
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 x 25 mm
50 x 50 mm
Catalog number 042820.FF
also known as 042820-FF
Price (EUR)/ Each
134,00
-
Add to cart
Quantity:
25 x 25 mm
Request bulk or custom format
Price (EUR)/ Each
134,00
Add to cart
Glassy carbon plate, 3mm (0.1 in.) thick, type 2, Thermo Scientific Chemicals
Catalog number042820.FF
Price (EUR)/ Each
134,00
-
Add to cart
Chemical Identifiers
CAS7440-44-0
Used in casting, smelting of metal, hard alloys. Glassy carbon is widely used as an electrode material in electrochemistry, for high temperature crucibles and as a part of some prosthetic devices.