Graphite foil, 0.4mm (0.015in) thick, 99.8% (metals basis), Thermo Scientific Chemicals
Graphite foil, 0.4mm (0.015in) thick, 99.8% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Graphite foil, 0.4mm (0.015in) thick, 99.8% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
15 x 15 cm
15 x 60 cm
15 x 250 cm
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 042952.VA
Price (EUR)/ Each
-
Quantity:
15 x 15 cm
Request bulk or custom format
Specifications
Chemical Name or MaterialGraphite
CAS7782-42-5
Recommended StorageAmbient temperatures
RTECS NumberMD9659600
EINECS Number231-955-3
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only