Thermo Scientific Chemicals

Tin(II) chloride dihydrate, ACS, 98.0-103.0%, Thermo Scientific Chemicals

Catalog number: 044409.36
500 g, Each
Thermo Scientific Chemicals

Tin(II) chloride dihydrate, ACS, 98.0-103.0%, Thermo Scientific Chemicals

Catalog number: 044409.36
500 g, Each
Quantity
Catalog number: 044409.36
also known as 044409-36
Price (EUR)
Quantity
-

Chemical Identifiers

CAS
10025-69-1
IUPAC Name
λ²-tin(2+) dihydrate dichloride
Molecular Formula
Cl2H4O2Sn
InChI Key
FWPIDFUJEMBDLS-UHFFFAOYSA-L
SMILES
O.O.[Cl-].[Cl-].[Sn++]
Impurity content
Na 0.01%
Impurity content
Pb 0.01%
Impurity content
SO₄ P.T.
Solution Test
in hydrochloric acid P.T.
Assay (unspecified)
98.0-103.0% Cl₂Sn.2H2O

Description

Stannous chloride is used as a sensitizing agent in preparing glass and plastic for metalizing. It serves as a potent reducing agent. Other uses of stannous chloride are tin electroplating baths, corrosion inhibitors, polymers, thermoplastic elastomers, soldering flux, antioxidant, tanning agent and pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text

    Safety and Handling


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Substances/mixtures corrosive to metal
    Category 1
    Acute oral toxicity
    Category 4
    Acute Inhalation Toxicity - Dusts and Mists
    Category 4
    Skin Corrosion/Irritation
    Category 1
    Serious Eye Damage/Eye Irritation
    Category 1
    Skin Sensitization
    Category 1
    Specific target organ toxicity - (single exposure)
    Category 3
    Specific target organ toxicity - (repeated exposure)
    Category 2
    Chronic aquatic toxicity
    Category 3
    Label Elements
    Signal Word

    Danger

    Hazard Statements

    H302 + H332 - Harmful if swallowed or if inhaled

    H314 - Causes severe skin burns and eye damage

    H317 - May cause an allergic skin reaction

    H335 - May cause respiratory irritation

    H373 - May cause damage to organs through prolonged or repeated exposure

    H412 - Harmful to aquatic life with long lasting effects

    Physical hazards

    H290 - May be corrosive to metals

    Precautionary Statements

    P280 - Wear protective gloves/protective clothing/eye protection/face protection

    P301 + P330 + P331 - IF SWALLOWED: Rinse mouth. Do NOT induce vomiting

    P303 + P361 + P353 - IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water or shower

    P304 + P340 - IF INHALED: Remove person to fresh air and keep comfortable for breathing

    P305 + P351 + P338 - IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing

    P310 - Immediately call a POISON CENTER or doctor/physician