Gadolinium powder, -200 mesh, 99.9% (REO), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Gadolinium powder, -200 mesh, 99.9% (REO), Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 045915.06
also known as 045915-06
Price (EUR)/ Each
122,40
Online exclusive
136.00 
Save 13,60 (10%)
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (EUR)/ Each
122,40
Online exclusive
136.00 
Save 13,60 (10%)
Add to cart
Gadolinium powder, -200 mesh, 99.9% (REO), Thermo Scientific Chemicals
Catalog number045915.06
Price (EUR)/ Each
122,40
Online exclusive
136.00 
Save 13,60 (10%)
-
Add to cart
Chemical Identifiers
CAS7440-54-2
IUPAC Namegadolinium
Molecular FormulaGd
InChI KeyUIWYJDYFSGRHKR-UHFFFAOYSA-N
SMILES[Gd]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.1% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Packaged under Argon
RUO – Research Use Only