Mebendazole
Mebendazole
Mebendazole
Thermo Scientific Chemicals

Mebendazole

CAS: 31431-39-7 | C16H13N3O3 | 295.30 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 101720250
Price (EUR)
94,35
Online Exclusive
111,00
Save 16,65 (15%)
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (EUR)
94,35
Online Exclusive
111,00
Save 16,65 (15%)
Each
Add to cart
Chemical Identifiers
CAS31431-39-7
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Appearance (Color)White to off-white to grey
Melting point285°C to 290°C
HPLC>=97.5 %
Loss on drying=<0.5 % (1 g, 105°C, 4 h)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Share catalog number, name or link