Thermo Scientific Chemicals

Diethyl 2-bromoethylphosphonate, 97%, Thermo Scientific Chemicals

Catalog number: 114180250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Diethyl 2-bromoethylphosphonate, 97%, Thermo Scientific Chemicals

Catalog number: 114180250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 114180250
Price (EUR)
Quantity
-

Chemical Identifiers

CAS
5324-30-1
IUPAC Name
diethyl (2-bromoethyl)phosphonate
Molecular Formula
C6H14BrO3P
InChI Key
PINITSMLVXAASM-UHFFFAOYSA-N
SMILES
CCOP(=O)(CCBr)OCC
Refractive index
1.4590 to 1.4610 (20°C, 589 nm)
Appearance (Color)
Clear colorless to light yellow
GC
>=96.0 %
Infrared spectrum
Conforms
Appearance (Form)
Liquid

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for C-C Bond Formation (Olefination)
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text

    Safety and Handling


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Skin Corrosion/Irritation
    Category 2
    Serious Eye Damage/Eye Irritation
    Category 2
    Specific target organ toxicity - (single exposure)
    Category 3
    Label Elements
    Signal Word

    Warning

    Hazard Statements

    H315 - Causes skin irritation

    H319 - Causes serious eye irritation

    H335 - May cause respiratory irritation

    Precautionary Statements

    P261 - Avoid breathing dust/fume/gas/mist/vapors/spray

    P280 - Wear protective gloves/protective clothing/eye protection/face protection

    P302 + P352 - IF ON SKIN: Wash with plenty of soap and water

    P305 + P351 + P338 - IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing