1x1 image pixel for data collection
4-Aminobenzotrifluoride, 99%
4-Aminobenzotrifluoride, 99%
4-Aminobenzotrifluoride, 99%
4-Aminobenzotrifluoride, 99%
Thermo Scientific Chemicals

4-Aminobenzotrifluoride, 99%

CAS: 455-14-1 | C7H6F3N | 161.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
10 mL
Packaging:
Glass bottle
Catalog number 208910500
Price (EUR)
159,80
Online Exclusive
188,00
Save 28,20 (15%)
Each
Add to cart
Quantity:
50 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
159,80
Online Exclusive
188,00
Save 28,20 (15%)
Each
Add to cart
Chemical Identifiers
IUPAC Name4-(trifluoromethyl)aniline
Molecular FormulaC7H6F3N
InChI KeyODGIMMLDVSWADK-UHFFFAOYSA-N
SMILESNC1=CC=C(C=C1)C(F)(F)F
Molecular Weight (g/mol)161.13
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to yellow to orange
Infrared spectrumConforms
GC>=98.5 %
Appearance (Form)Liquid
Refractive index1.4830 to 1.4860 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Share catalog number, name or link