Chloral hydrate, 98.5%
Product Image
Thermo Scientific Chemicals

Chloral hydrate, 98.5%

CAS: 302-17-0 | C2H3Cl3O2 | 165.39 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
500 g
2.5 kg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 232470500
Price (EUR)
24,57
Online Exclusive
29,60
Save 5,03 (17%)
Each
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (EUR)
24,57
Online Exclusive
29,60
Save 5,03 (17%)
Each
Add to cart
Chemical Identifiers
CAS302-17-0
IUPAC Name2,2,2-trichloroethane-1,1-diol
Molecular FormulaC2H3Cl3O2
InChI KeyRNFNDJAIBTYOQL-UHFFFAOYSA-N
SMILESOC(O)C(Cl)(Cl)Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to white
Appearance (Form)Powder or crystals
Infrared spectrumConforms
Titration excess NaOH98.0 to 102.0 %
Chloride (Cl)=<200 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Please enter sku to load default questions

Documents & Downloads

Certificates

Safety Data Sheets

Safety and Handling

Classification of the substance or mixture
CLP classification - Regulation(EC) No 1272/2008
Acute oral toxicity
Category 3
Skin Corrosion/Irritation
Category 2
Serious Eye Damage/Eye Irritation
Category 2
Label Elements
Signal Word

Danger

Hazard Statements

H301 - Toxic if swallowed

H315 - Causes skin irritation

H319 - Causes serious eye irritation

Precautionary Statements

P280 - Wear protective gloves/protective clothing/eye protection/face protection

P302 + P352 - IF ON SKIN: Wash with plenty of soap and water

P305 + P351 + P338 - IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing

P301 + P310 - IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician

Share catalog number, name or link