L-alpha-Dipalmitoyl phosphatidylcholine, 98%
L-alpha-Dipalmitoyl phosphatidylcholine, 98%
L-alpha-Dipalmitoyl phosphatidylcholine, 98%
L-alpha-Dipalmitoyl phosphatidylcholine, 98%
Thermo Scientific Chemicals

L-alpha-Dipalmitoyl phosphatidylcholine, 98%

CAS: 63-89-8 | C40H80NO8P | 734.053 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mg
500 mg
2.5 g
Catalog number 240571000
Price (EUR)
82,37
Online Exclusive
96,90
Save 14,53 (15%)
Each
Estimated availability date  Pending  
Add to cart
Quantity:
100 mg
Request bulk or custom format
Price (EUR)
82,37
Online Exclusive
96,90
Save 14,53 (15%)
Each
Add to cart
Chemical Identifiers
CAS63-89-8
IUPAC Name(2-{[2,3-bis(hexadecanoyloxy)propyl phosphonato]oxy}ethyl)trimethylazanium
Molecular FormulaC40H80NO8P
InChI KeyKILNVBDSWZSGLL-UHFFFAOYNA-N
SMILESCCCCCCCCCCCCCCCC(=O)OCC(COP([O-])(=O)OCC[N+](C)(C)C)OC(=O)CCCCCCCCCCCCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Infrared spectrumConforms
Specific optical rotation+6° to +8° (20°C, 589 nm) (c=2, CHCl3)
Titration with HClO4>=97.5 %
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0470224Certificate of AnalysisMay 17, 2025240570025, 240571000, 240575000
A0452065Certificate of AnalysisMay 02, 2023240570025, 240571000, 240575000
A0440636Certificate of AnalysisMar 31, 2022240570025, 240571000, 240575000
A0425434Certificate of AnalysisJan 06, 2021240570025, 240571000, 240575000
A0404421Certificate of AnalysisOct 01, 2020240570025, 240571000, 240575000
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection