Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: LLCKRCWDVH QRLNRAMEEA EKTTTTYLDN GTHPAQDPDF RGEDPECQDA ETERFLSTSS TGRRVSFNEA ALFEQSRKTQ DKGRRYTLTE GDFHHLKNAR LTHLHLPPLK IVTIHECDSG EASSATTPHP ATSPKATLAI FQPPGKALTG RSVGPSSALP GDPYNSAAGA TDFAEISPSA SSDSGEGTSL DAGTRSTKAG GP
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: calcium channel, voltage-dependent, beta subunit associated regulatory protein; downstream of Stk11; protein Dos; VGCC beta-anchoring and -regulatory protein; Voltage-dependent calcium channel beta subunit-associated regulatory protein
Gene Aliases: BARP; C19orf26; CBARP; DOS
UniProt ID: (Human) Q8N350
Entrez Gene ID: (Human) 255057
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support