4-Isopropylbenzoic acid, 98+%, Thermo Scientific Chemicals
4-Isopropylbenzoic acid, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Isopropylbenzoic acid, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number 412800250
Price (EUR)/ Each
127,80
Online exclusive
142,00 
Save 14,20 (10%)
-
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)/ Each
127,80
Online exclusive
142,00 
Save 14,20 (10%)
Add to cart
4-Isopropylbenzoic acid, 98+%, Thermo Scientific Chemicals
Catalog number412800250
Price (EUR)/ Each
127,80
Online exclusive
142,00 
Save 14,20 (10%)
-
Add to cart
Chemical Identifiers
IUPAC Name4-(propan-2-yl)benzoic acid
Molecular FormulaC10H12O2
InChI KeyCKMXAIVXVKGGFM-UHFFFAOYSA-N
SMILESCC(C)C1=CC=C(C=C1)C(O)=O
Molecular Weight (g/mol)164.20
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point116°C to 120°C
HPLC>=98.0 %
Appearance (Color)White
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only