Ethosuximide
Ethosuximide
Ethosuximide
Thermo Scientific Chemicals

Ethosuximide

CAS: 77-67-8 | C7H11NO2 | 141.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number 461401000
Price (EUR)
277,10
Precio exclusivo en nuestra web
326,00
Save 48,90 (15%)
Each
-
Add to cart
Quantity:
100 g
Recurring order eligible. Learn more »
Request bulk or custom format
Price (EUR)
277,10
Precio exclusivo en nuestra web
326,00
Save 48,90 (15%)
Each
Add to cart
Chemical Identifiers
CAS77-67-8
IUPAC Name3-ethyl-3-methylpyrrolidine-2,5-dione
Molecular FormulaC7H11NO2
InChI KeyHAPOVYFOVVWLRS-UHFFFAOYNA-N
SMILESCCC1(C)CC(=O)NC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or chunks
Water=<0.5 %
Infrared spectrumConforms
Melting point47°C to 52°C
HPLC>=97.5 %
View more
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0375201Certificate of AnalysisMar 08, 2024461401000, 461400250, 461400050
1 result displayed, search above for a specific certificate

Safety Data Sheets

Safety and Handling

Classification of the substance or mixture
CLP classification - Regulation(EC) No 1272/2008
Acute oral toxicity
Category 4
Germ Cell Mutagenicity
Category 2
Reproductive Toxicity
Category 1A
Label Elements
Signal Word

Danger

Hazard Statements

H302 - Harmful if swallowed

H341 - Suspected of causing genetic defects

H360D - May damage the unborn child

Precautionary Statements

P201 - Obtain special instructions before use

P264 - Wash face, hands and any exposed skin thoroughly after handling

P280 - Wear protective gloves/protective clothing/eye protection/face protection

P301 + P330 + P331 - IF SWALLOWED: rinse mouth. Do NOT induce vomiting

P312 - Call a POISON CENTER or doctor if you feel unwell

Additional EU labelling

Restricted to professional users

Share catalog number, name or link