1x1 image pixel for data collection
Chlorothiazide, 98%
Chlorothiazide, 98%
Chlorothiazide, 98%
Thermo Scientific Chemicals

Chlorothiazide, 98%

CAS: 58-94-6 | C7H6ClN3O4S2 | 295.71 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number 461741000
Price (EUR)
192,10
Online Exclusive
232,00
Save 39,90 (17%)
Each
Estimated availability date  Pending  
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (EUR)
192,10
Online Exclusive
232,00
Save 39,90 (17%)
Each
Add to cart
Chemical Identifiers
CAS58-94-6
SpecificationsSpecification SheetSpecification Sheet
HPLC>=97.5 %
Appearance (Form)Powder
Appearance (Color)White to off-white
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Safety and Handling

Classification of the substance or mixture
CLP classification - Regulation(EC) No 1272/2008
Respiratory Sensitization
Category 1
Skin Sensitization
Category 1
Label Elements
Signal Word

Danger

Hazard Statements

H317 - May cause an allergic skin reaction

H334 - May cause allergy or asthma symptoms or breathing difficulties if inhaled

Precautionary Statements

P280 - Wear protective gloves/protective clothing

P285 - In case of inadequate ventilation wear respiratory protection

P302 + P352 - IF ON SKIN: Wash with plenty of soap and water

P304 + P340 - IF INHALED: Remove person to fresh air and keep comfortable for breathing

P333 + P313 - If skin irritation or rash occurs: Get medical advice/attention

P342 + P311 - If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician

Share catalog number, name or link

1x1 image pixel for data collection