1x1 image pixel for data collection
Clofazimine, 98%
Clofazimine, 98%
Clofazimine, 98%
Thermo Scientific Chemicals

Clofazimine, 98%

CAS: 2030-63-9 | C27H22Cl2N4 | 473.4 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 461760010
Price (EUR)
52,11
Online Exclusive
62,80
Save 10,69 (17%)
Each
Estimated availability date  Pending  
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (EUR)
52,11
Online Exclusive
62,80
Save 10,69 (17%)
Each
Add to cart
Chemical Identifiers
CAS2030-63-9
IUPAC NameN,5-bis(4-chlorophenyl)-3-[(propan-2-yl)imino]-3,5-dihydrophenazin-2-amine
Molecular FormulaC27H22Cl2N4
InChI KeyWDQPAMHFFCXSNU-UHFFFAOYSA-N
SMILESCC(C)N=C1C=C2N(C3=CC=C(Cl)C=C3)C3=CC=CC=C3N=C2C=C1NC1=CC=C(Cl)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Loss on drying=<0.5 % (105°C)
Appearance (Color)Orange to red to brown
Infrared spectrumConforms
Titration with HClO4>=97.5 %
HPLC>=97.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection