Sumatriptan succinate
Sumatriptan succinate
Thermo Scientific Chemicals

Sumatriptan succinate

CAS: 103628-48-4 | C18H27N3O6S | 413.49 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 462420010
Price (EUR)
157,25
Online Exclusive
190,00
Save 32,75 (17%)
Each
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (EUR)
157,25
Online Exclusive
190,00
Save 32,75 (17%)
Each
Add to cart
Chemical Identifiers
CAS103628-48-4
IUPAC Name1-{3-[2-(dimethylamino)ethyl]-1H-indol-5-yl}-N-methylmethanesulfonamide; butanedioic acid
Molecular FormulaC18H27N3O6S
InChI KeyPORMUFZNYQJOEI-UHFFFAOYSA-N
SMILESOC(=O)CCC(O)=O.CNS(=O)(=O)CC1=CC=C2NC=C(CCN(C)C)C2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Water=<1 %
Infrared spectrumConforms
Melting point165°C to 169°C
HPLC>=97.5 %
Solubility(2.5 % in water) Clear colorless solution
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Please enter sku to load default questions

Documents & Downloads

Certificates

Safety Data Sheets

Safety and Handling

Classification of the substance or mixture
CLP classification - Regulation(EC) No 1272/2008
Serious Eye Damage/Eye Irritation
Category 1
Label Elements
Signal Word

Danger

Hazard Statements

H318 - Causes serious eye damage

Precautionary Statements

P280 - Wear eye protection/ face protection

P305 + P351 + P338 - IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing

P310 - Immediately call a POISON CENTER or doctor/physician

Share catalog number, name or link