Betamethasone sodium phosphate
Betamethasone sodium phosphate
Thermo Scientific Chemicals

Betamethasone sodium phosphate

Have Questions?
Catalog NumberQuantity
4657400505 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 465740050
Price (EUR)
238,00
Online Exclusive
287,00
Save 49,00 (17%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (EUR)
238,00
Online Exclusive
287,00
Save 49,00 (17%)
Each
Add to cart
Chemical Identifiers
CAS151-73-5
IUPAC Namedisodium (1R,2S,3aS,3bS,9aS,9bR,10S,11aS)-9b-fluoro-1,10-dihydroxy-2,9a,11a-trimethyl-1-[2-(phosphonatooxy)acetyl]-1H,2H,3H,3aH,3bH,4H,5H,7H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-7-one
Molecular FormulaC22H28FNa2O8P
InChI KeyPLCQGRYPOISRTQ-LWCNAHDDSA-L
SMILES[Na+].[Na+].C[C@H]1C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]2(C)[C@@]1(O)C(=O)COP([O-])([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
HPLC>=96.0 %
Appearance (Form)Powder
Specific optical rotation+98° to +105° (20°C, 589 nm) (1% in water)
Appearance (Color)White to almost white
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Safety and Handling

Classification of the substance or mixture
CLP classification - Regulation(EC) No 1272/2008
Acute oral toxicity
Category 4
Reproductive Toxicity
Category 2
Label Elements
Signal Word

Warning

Hazard Statements

H302 - Harmful if swallowed

H361 - Suspected of damaging fertility or the unborn child

Precautionary Statements

P201 - Obtain special instructions before use

P264 - Wash face, hands and any exposed skin thoroughly after handling

P280 - Wear protective gloves/protective clothing/eye protection/face protection

P301 + P330 + P331 - IF SWALLOWED: rinse mouth. Do NOT induce vomiting

P312 - Call a POISON CENTER or doctor if you feel unwell

Share catalog number, name or link