Tazarotene
Thermo Scientific Chemicals

Tazarotene

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 466300010
Price (EUR)
685,95
Precio exclusivo en nuestra web
827,00
Save 141,05 (17%)
Each
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (EUR)
685,95
Precio exclusivo en nuestra web
827,00
Save 141,05 (17%)
Each
Add to cart
Chemical Identifiers
CAS118292-40-3
IUPAC Nameethyl 6-[2-(4,4-dimethyl-3,4-dihydro-2H-1-benzothiopyran-6-yl)ethynyl]pyridine-3-carboxylate
Molecular FormulaC21H21NO2S
InChI KeyOGQICQVSFDPSEI-UHFFFAOYSA-N
SMILESCCOC(=O)C1=CC=C(N=C1)C#CC1=CC=C2SCCC(C)(C)C2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Loss on drying=<1 %
HPLC>=97.5 %
Melting point102°C to 106°C
Appearance (Color)White to light yellow
Appearance (Form)Crystalline powder
View more
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Kunden, die diesen Artikel ansahen, interessierten sich auch für



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0418272Certificate of AnalysisMay 17, 2025466300010, 466302500
A0418272Certificate of AnalysisNov 07, 2022466300010, 466302500
2 results displayed, search above for a specific certificate

Safety Data Sheets

Safety and Handling

Classification of the substance or mixture
CLP classification - Regulation(EC) No 1272/2008
Skin Corrosion/Irritation
Category 2
Serious Eye Damage/Eye Irritation
Category 2
Specific target organ toxicity - (single exposure)
Category 3
Label Elements
Signal Word

Warning

Hazard Statements

H315 - Causes skin irritation

H319 - Causes serious eye irritation

H335 - May cause respiratory irritation

Precautionary Statements

P280 - Wear protective gloves/protective clothing/eye protection/face protection

P302 + P352 - IF ON SKIN: Wash with plenty of soap and water

P304 + P340 - IF INHALED: Remove person to fresh air and keep comfortable for breathing

P305 + P351 + P338 - IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing

P312 - Call a POISON CENTER or doctor if you feel unwell

Share catalog number, name or link