Zopiclone
Thermo Scientific Chemicals

Zopiclone

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number 467232500
Price (EUR)
111,35
Online Exclusive
131,00
Save 19,65 (15%)
Each
-
Add to cart
Quantity:
250 mg
Request bulk or custom format
Price (EUR)
111,35
Online Exclusive
131,00
Save 19,65 (15%)
Each
Add to cart
Chemical Identifiers
CAS43200-80-2
SpecificationsSpecification SheetSpecification Sheet
Loss on drying=<0.5 % (105°C, 2 hr)
HPLC>=98.5 %
Appearance (Color)White to light yellow
Appearance (Form)Powder
Infrared spectrumConforms
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0420448Certificate of AnalysisJan 28, 2025467230010, 467232500
A0420448Certificate of AnalysisAug 25, 2023467230010, 467232500
2 results displayed, search above for a specific certificate

Safety Data Sheets

Safety and Handling

Classification of the substance or mixture
CLP classification - Regulation(EC) No 1272/2008
Acute oral toxicity
Category 4
Acute dermal toxicity
Category 4
Acute Inhalation Toxicity - Dusts and Mists
Category 4
Skin Corrosion/Irritation
Category 2
Serious Eye Damage/Eye Irritation
Category 2
Reproductive Toxicity
Category 2
Specific target organ toxicity - (single exposure)
Category 3
Label Elements
Signal Word

Warning

Hazard Statements

H302 + H312 + H332 - Harmful if swallowed, in contact with skin or if inhaled

H315 - Causes skin irritation

H319 - Causes serious eye irritation

H335 - May cause respiratory irritation

H361 - Suspected of damaging fertility or the unborn child

Precautionary Statements

P280 - Wear protective gloves/protective clothing/eye protection/face protection

P301 + P330 + P331 - IF SWALLOWED: rinse mouth. Do NOT induce vomiting

P302 + P352 - IF ON SKIN: Wash with plenty of soap and water

P304 + P340 - IF INHALED: Remove person to fresh air and keep comfortable for breathing

P305 + P351 + P338 - IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing

P312 - Call a POISON CENTER or doctor if you feel unwell

Share catalog number, name or link

1x1 image pixel for data collection