L-Erythrulose
Thermo Scientific Chemicals

L-Erythrulose

CAS: 533-50-6 | C4H8O4, C4H8O4 | 120.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 467301000
Price (EUR)
193,80
キャンペーン価格
228,00
Save 34,20 (15%)
Each
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (EUR)
193,80
キャンペーン価格
228,00
Save 34,20 (15%)
Each
Add to cart
Chemical Identifiers
CAS533-50-6
IUPAC Name(3S)-1,3,4-trihydroxybutan-2-one
Molecular FormulaC4H8O4
InChI KeyUQPHVQVXLPRNCX-YEQUQEJONA-N
SMILESOC[C@H](O)C(=O)CO
View more
SpecificationsSpecification SheetSpecification Sheet
Water=<24.0 %
Infrared spectrumConforms
Appearance (Color)Yellow to green to brown
HPLC>=76.0 %
Sulfated ash=<4.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Share catalog number, name or link