Digital Analog Converter Boards for UltiMate™ 3000 Optical Detector Upgrades
Digital Analog Converter Boards for UltiMate™ 3000 Optical Detector Upgrades
Actual product may vary
Thermo Scientific™
Digital Analog Converter Boards for UltiMate™ 3000 Optical Detector Upgrades
Digital Analog Converter Boards for UltiMate 3000 Optical Detector Upgrades provide an optional means for wavelength detectors to yield analog output.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Equipment)
6078.0305FLD-3000 Series
6074.0305VWD-3000 Series
6082.0305DAD-3000 Series, MWD-3000 Series
Catalog number 6078.0305
Price (EUR)/ Each
-
Request A Quote
For Use With (Equipment):
FLD-3000 Series
Provide an optional means for wavelength detectors to yield analog output with Thermo Scientific™ Dionex Digital Analog Converter Boards for UltiMate™ 3000 Optical Detector Upgrades.
  • Two analog outputs
  • Configurable for use with Chromeleon™ software
Specifications
DescriptionDAC Board for FLD-3x00(RS)
For Use With (Equipment)FLD-3000 Series
Product LineUltiMate
TypeInterface Accessories
FrequencyUp to 50 Hz
Unit SizeEach