Thermo Scientific Chemicals

L-Phenylalaninol, 98%, Thermo Scientific Chemicals

Catalog number: A11586.03
1 g, Each
Thermo Scientific Chemicals

L-Phenylalaninol, 98%, Thermo Scientific Chemicals

Catalog number: A11586.03
1 g, Each
Quantity
Catalog number: A11586.03
also known as A11586-03
Price (EUR)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3182-95-4
IUPAC Name
(2S)-2-amino-3-phenylpropan-1-ol
Molecular Formula
C9H13NO
InChI Key
STVVMTBJNDTZBF-VIFPVBQESA-N
SMILES
N[C@H](CO)CC1=CC=CC=C1
Melting Point (clear melt)
88.0-94.0°C
Optical Rotation
-23.3 to -25.3° (C=5 in ethanol)
Appearance (Color)
White to cream to yellow
Form
Crystals or powder or crystalline powder
Assay (Non-aqueous acid-base Titration)
≥97.5 to ≤102.5%

Description

L-Phenylalaninol acts as an antiulcer agent that reduces the secretion of gastric acid and prevents ulcer formation. It inhibits intestinal absorption of L-phenylalanine making it a prospective treatment for phenylketonuria. Further, it reacts with nitriles to form oxazolines, which finds application in palladium-catalyzed allylic substitution. In addition to this, it is also used in amidation for chiral resolution and nicotinamide adenine dinucleotide modeling.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Phenylalaninol acts as an antiulcer agent that reduces the secretion of gastric acid and prevents ulcer formation. It inhibits intestinal absorption of L-phenylalanine making it a prospective treatment for phenylketonuria. Further, it reacts with nitriles to form oxazolines, which finds application in palladium-catalyzed allylic substitution. In addition to this, it is also used in amidation for chiral resolution and nicotinamide adenine dinucleotide modeling.

Solubility
Soluble in chloroform, ethyl acetate, ethanol and methanol.

Notes
Incompatible with acids, acid chlorides, acid anhydrides and oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text

    Safety and Handling


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Skin Corrosion/Irritation
    Category 1
    Serious Eye Damage/Eye Irritation
    Category 1
    Label Elements
    Signal Word

    Danger

    Hazard Statements

    H314 - Causes severe skin burns and eye damage

    Precautionary Statements

    P280 - Wear eye protection/ face protection

    P301 + P330 + P331 - IF SWALLOWED: rinse mouth. Do NOT induce vomiting

    P305 + P351 + P338 - IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing

    P310 - Immediately call a POISON CENTER or doctor/physician