Thermo Scientific Chemicals

Isopropyl isothiocyanate, 97%, Thermo Scientific Chemicals

Catalog number: A11641.14
25 g, Each
Thermo Scientific Chemicals

Isopropyl isothiocyanate, 97%, Thermo Scientific Chemicals

Catalog number: A11641.14
25 g, Each
Quantity
Catalog number: A11641.14
also known as A11641-14
Price (EUR)
Quantity
-

Chemical Identifiers

CAS
2253-73-8
IUPAC Name
2-isothiocyanatopropane
Molecular Formula
C4H7NS
InChI Key
VHBFEIBMZHEWSX-UHFFFAOYSA-N
SMILES
CC(C)N=C=S
Form
Liquid
Appearance (Color)
Clear colorless to yellow
Assay (GC)
≥96.0%
Identification (FTIR)
Conforms
Refractive Index
1.4910-1.4960 @ 20°C

Description

Isopropyl isothiocyanate is used as is an antimicrobial agent. It is also used as medicinal intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Isopropyl isothiocyanate is used as is an antimicrobial agent. It is also used as medicinal intermediate.

Solubility
Miscible with methanol. Hydrolyzes in water.

Notes
Moisture Sensitive. Protect from humidity and water. Store under dry inert gas away from oxidizing agents and moisture. Incompatible with alcohols and amines.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text

    Safety and Handling


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Flammable liquids
    Category 3
    Skin Corrosion/Irritation
    Category 2
    Serious Eye Damage/Eye Irritation
    Category 2
    Specific target organ toxicity - (single exposure)
    Category 3
    Label Elements
    Signal Word

    Warning

    Hazard Statements

    H315 - Causes skin irritation

    H319 - Causes serious eye irritation

    H335 - May cause respiratory irritation

    Physical Hazards

    H226 - Flammable liquid and vapor

    Precautionary Statements

    P210 - Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking

    P280 - Wear protective gloves/protective clothing/eye protection/face protection

    P303 + P361 + P353 - IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water or shower

    P312 - Call a POISON CENTER or doctor if you feel unwell

    P332 + P313 - If skin irritation occurs: Get medical advice/attention

    P337 + P313 - If eye irritation persists: Get medical advice/attention