Thermo Scientific Chemicals

2-Pyridineacetonitrile, 97%, Thermo Scientific Chemicals

Catalog number: A13087.14
25 g, Each
Thermo Scientific Chemicals

2-Pyridineacetonitrile, 97%, Thermo Scientific Chemicals

Catalog number: A13087.14
25 g, Each
Quantity
Catalog number: A13087.14
also known as A13087-14
Price (EUR)
Quantity
-

Chemical Identifiers

CAS
2739-97-1
IUPAC Name
2-(pyridin-2-yl)acetonitrile
Molecular Formula
C7H6N2
InChI Key
UKVQBONVSSLJBB-UHFFFAOYSA-N
SMILES
N#CCC1=CC=CC=N1
Appearance (Color)
White to yellow to orange to red to dark brown
Refractive Index
1.5230-1.5300 as melt
Form
Low melting solid (clear liquid as melt)
Assay (GC)
≥96.5%

Description

2-Pyridineacetonitrile is used as a pharmaceutical intermediate. It is also used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Pyridineacetonitrile is used as a pharmaceutical intermediate. It is also used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Miscible with water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents, strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text

    Safety and Handling


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Acute oral toxicity
    Category 3
    Acute dermal toxicity
    Category 2
    Skin Corrosion/Irritation
    Category 2
    Serious Eye Damage/Eye Irritation
    Category 2
    Specific target organ toxicity - (single exposure)
    Category 3
    Label Elements
    Signal Word

    Danger

    Hazard Statements

    H301 - Toxic if swallowed

    H310 - Fatal in contact with skin

    H315 - Causes skin irritation

    H319 - Causes serious eye irritation

    H335 - May cause respiratory irritation

    EU Specific Hazard Statements

    Combustible liquid

    Precautionary Statements

    P280 - Wear protective gloves/protective clothing/eye protection/face protection

    P301 + P330 + P331 - IF SWALLOWED: rinse mouth. Do NOT induce vomiting

    P302 + P352 - IF ON SKIN: Wash with plenty of soap and water

    P304 + P312 - IF INHALED: Call a POISON CENTER or doctor if you feel unwell

    P310 - Immediately call a POISON CENTER or doctor/physician

    P337 + P313 - If eye irritation persists: Get medical advice/attention